Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad50 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Rad50 |
---|---|
Dilution | Western Blot 1:2000, Simple Western 1:2000, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 1:10-1:500, ICC/IF (Negative) |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100154SS
![]() |
Novus Biologicals
NB100154SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB100154
![]() |
Novus Biologicals
NB100154 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Rad50 Polyclonal specifically detects Rad50 in Human, Mouse, Hamster samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, ICC/IF (Negative).Specifications
Rad50 | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining | |
Whole antisera with 0.02% Sodium Azide | |
10111 | |
C-terminal mouse RAD50 (604 amino acids). [UniProt# P70388] | |
Primary | |
Store at 4C. Do not freeze. |
Western Blot 1:2000, Simple Western 1:2000, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 1:10-1:500, ICC/IF (Negative) | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Hamster | |
DNA repair protein RAD50, EC 3.6, EC 3.6.1.15, EC 3.6.3, EC 3.6.3.27, hRAD50, NBSLD, RAD50 (S. cerevisiae) homolog, RAD50 homolog (S. cerevisiae), RAD502, RAD50-2 | |
RAD50 | |
IgG | |
Unpurified | |
153 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title