Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAG1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

$206.00 - $548.00

Specifications

Antigen RAG1
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17419020
SDP
View Documents
Novus Biologicals
NBP17419020UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP174190
SDP
View Documents
Novus Biologicals
NBP174190
100 μL
Each for $548.00
Only null left
Add to Cart
 
Description

Description

RAG1 Polyclonal specifically detects RAG1 in Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Specifications

Specifications

RAG1
Unconjugated
RUO
MGC43321, RAG-1, recombination activating gene 1, RING finger protein 74, RNF74recombination activating protein 1, V(D)J recombination-activating protein 1
RAG1
IgG
119 kDa
Polyclonal
Rabbit
P15919
5896
Synthetic peptides corresponding to the C terminal of Rag1. Immunizing peptide sequence IIERDGSIGAWASEGNESGNKLFRRFRKMNARQSKCYEMEDVLKHHWLYT.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.