Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ RapidScript™ Probe One-Step RT-qPCR Kit

Catalog No. mp116188000 Shop All MP Biomedicals, Inc Products
Click to view available options
:
No. of Reactions: 100

Use the RapidScript™ Probe One-Step RT-qPCR Kit for simple, one-step RT-qPCR.

The RapidScript™ Probe One-Step RT-qPCR Kit provides the convenience of an all-in-one kit through the combination of an RT enzyme mix and a 2X qPCR mastermix to create a one-step reverse transcription and real-time qPCR amplification, all in one PCR reaction tube.

The unique formulation of the RapidScript™ RT-qPCR enzyme allows the reaction to be run either at standard conditions or fast conditions, depending on the capacity of the qPCR instrument. This allows for the RT-qPCR reaction to be completed in a total of ∼68 minutes (using the standard protocol) or ∼24 minutes (using the Fast protocol), as opposed to conventional reactions with over 50 minutes for the RT reaction and 1.5-2 hours for qPCR amplification.

  • Ease of use, user flexibility and time savings when using Standard or Fast protocols (compared to conventional two-step RT and qPCR reactions)
  • The mastermix is suitable for SNP genotyping assays, gene expression analysis, microarray validation and high throughput screening, among others
  • ROX reference dye is provided separately, allowing for the reaction to be universally compatible with most qPCR instruments
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2X qPCR MasterMix, 1.25 mL; RT-qPCR Enzyme Mix, 40 μL; ROX Reerence Dye, 15 μL; 10 mM Oligo(dT), 100 μL. Store at -20°C
Format Kit
Polymerase RapidScript RT-qPCR enzyme
Reaction Speed ∼68 min. (Standard)/∼24 min. (Fast)
Passive Reference Dye ROX
Reverse Transcriptase Yes
For Use With (Application) Gene Expression
No. of Reactions 100

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.