Learn More
GL7 (T and B Cell Activation Marker) Antibody (GL-7), Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | GL7 (T and B Cell Activation Marker) |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | GL-7 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry < = 0.5 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Formulation | PBS, pH 7.2 with 0.09% Sodium Azide |
| Host Species | Rat |
| Immunogen | The immunogen for this antibody is T- and B-cell activation antigen (GL7, Ly-77). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.