Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat anti-MHC Class II, Clone: M5/114.15.2, PE, Novus Biologicals™

Rat Monoclonal Antibody
Specifications
| Antigen | MHC Class II |
|---|---|
| Clone | M5/114.15.2 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000 |
| Applications | Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14376705
![]() |
Novus Biologicals
NBP1437670.05MG |
0.05 mg | N/A | N/A | N/A | ||||
NBP14376701
![]() |
Novus Biologicals
NBP1437670.1MG |
0.1 mg | N/A | N/A | N/A | ||||
NBP14376702
![]() |
Novus Biologicals
NBP1437670.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
MHC class II (I-A/I-E) Monoclonal specifically detects MHC class II (I-A/I-E) in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| MHC Class II | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Mouse | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| HLA-DMA | |
| IgG2b κ | |
| Affinity Purified | |
| The monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determinant present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes. |
| M5/114.15.2 | |
| Flow Cytometry 1:10-1:1000 | |
| Monoclonal | |
| Rat | |
| Adaptive Immunity, Immune System Diseases, Immunology, Major Histocompatibility Complexes, Transcription Factors and Regulators | |
| Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
| 3108 | |
| The immunogen for this antibody was MHC Class II. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title