Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Raw Concrete Pack
SCI_ED

Catalog No. S012307 Shop All Eisco Products
Click to view available options
Grade:
9 to 12

Catalog No. S012307

Supplier: Eisco™ ESNG0089PK12

Only null left
Add to Cart
Add to Cart

A fun introduction to the world of geology through a 12 piece set. Each Tech Cut specimen is selected and hand processed by an actual geologist, ensuring each sample exhibits defining features.

  • While each specimen will vary in shape and size, they will measure approximately 1 in. each
  • Test hardness, streak color, magnetic properties, appearance, and effervescence to identify the different specimens in this kit

Specifications

Product Type Basic Rocks and Minerals Kit
Description 12 piece
For Use With (Application) For demonstration
Grade 9 to 12
Quantity 12
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.