Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBMS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | RBMS1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15823220
![]() |
Novus Biologicals
NBP15823220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP158232
![]() |
Novus Biologicals
NBP158232 |
100 μL |
Each for $487.50
|
|
|||||
Description
RBMS1 Polyclonal specifically detects RBMS1 in Human samples. It is validated for Western Blot.Specifications
RBMS1 | |
Polyclonal | |
Purified | |
RUO | |
chromosome 2 open reading frame 12, DKFZp564H0764, HCC-4, MGC15146, MGC3331, MGC97258, MGC97270, MGC97282, MGC99543, MSSP1, MSSP-1, MSSP-2, MSSP-3, RNA binding motif, single stranded interacting protein 1, SCR2MGC70597, single-stranded-interacting protein 1, suppressor of cdc 2 (cdc13) with RNA binding motif 2 | |
RBMS1 | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
P29558-2 | |
5937 | |
Synthetic peptides corresponding to RBMS1 (RNA binding motif, single stranded interacting protein 1) The peptide sequence was selected from the middle region of RBMS1. Peptide sequence GVSAPTEPLLCKFADGGQKKRQNPNKYIPNGRPWHREGEAGMTLTYDPTT. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title