Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Reagent Powder for CellTiter 96™ AQueous Nonradioactive Cell Proliferation Assay Kit (MTS)
Shop All Promega Corporation Products
Click to view available options
Quantity:
250 mg
1 g
Description
- A Novel Tetrazolium Compound for Use in Colorimetric Assays
- Available in two sizes
- Use in assays to determine the number of viable cells
Specifications
Specifications
Quantity | 250 mg |
Detection Method | Colorimetric |
For Use With (Application) | Determining the number of viable cells in proliferation |
Content And Storage | -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction