Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Red Sandstone Specimen
Shop All Eisco Products

Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Description
- Color of this sample is due to oxidized iron becoming interspersed with sand grains during lithification
- For use in comparing white sandstone to understand physical expression of chemical differences
- Correlates well to Next Generation Science Standards Evidence Statements
- Red color due to inclusion of iron in chemical structure of rock
Specifications
Specifications
| Product Type | Sedimentary Rock, Red Sandstone |
| For Use With (Application) | Comparing white sandstone to understand physical expression of chemical differences |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction