Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reduced Folate Carrier/SLC19A1 Antibody (890513), HRP, Novus Biologicals™
SDP

Catalog No. NB346086 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346086 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346086 Supplier Novus Biologicals Supplier No. FAB8450H
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reduced Folate Carrier/SLC19A1 Monoclonal antibody specifically detects Reduced Folate Carrier/SLC19A1 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Reduced Folate Carrier/SLC19A1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 890513
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias CHMD, FLOT1, folate transporter 1, FOLThuman reduced folate carrier (RFC)10Intestinal folate carrier 1, IFC1, IFC-1, Placental folate transporter, Reduced folate carrier protein, REFC, RFC, RFC1, solute carrier family 19 (folate transporter), member 1, Solute carrier family 19 member 1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Reduced Folate Carrier/SLC19A1, Met1-Gln591, Accession # P41440
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6573
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.