Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ RelA/NFkB p65 [p Ser276] Inhibitor Peptide Set
SDP

Catalog No. p-5538754 Shop All R&D Systems Products
Click to view available options
Quantity:
2 mg
5 mg

Applications: In vivo assay, Functional (Inhibition), Block/Neutralize

Specifications

Host Species Human, Mouse, Rat, Bovine, Canine, Mammal
Components NF-kB p65 (Ser276) Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKQLRRPSDRELSE (p65 sequence: QLRRPSDRELSE). Molecular weight: 3830; Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
For Use With (Application) Blocking/Neutralizing, Functional (Inhibition), In vivo assay
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 5 mg
Product Type RelA/NFkB p65 [p Ser276] Inhibitor Peptide Set
Inhibitors RelA/NFkB p65 [p Ser276]
Form Lyophilized

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.