Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Renal Cell Carcinoma Marker (gp200) Ab-1 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Renal Cell Carcinoma Marker (gp200) Ab-1, Mouse Monoclonal Antibody.

$203.00 - $963.00

Specifications

Antigen Renal Cell Carcinoma Marker (gp200) Ab-1
Clone PN-15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Mouse
View More Specs

Products 9
Catalog Number Mfr. No. Form Conjugate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Conjugate Quantity Price Quantity & Availability  
MS409RQ
View Documents
Epredia
MS409RQ
Ready-to-Use Unconjugated 12 mL
Each for $559.00
Only null left
Add to Cart
 
MS409P0
View Documents
Epredia
MS409P0
Concentrated Unconjugated 100 μL
Each for $203.00
Only null left
Add to Cart
 
MS409P
View Documents
Epredia
MS409P
Concentrated Unconjugated 1 mL
Each for $963.00
Only null left
Add to Cart
 
MS409P1ABX
View Documents
Epredia
MS409P1ABX
Concentrated Unconjugated 100 μL
Each for $602.00
Only null left
Add to Cart
 
MS409PABX
View Documents
Epredia
MS409PABX
Concentrated Unconjugated 200 μL
Each for $802.00
Only null left
Add to Cart
 
MS409R7
View Documents
Epredia
MS409R7
Prediluted Unconjugated 7 mL
Each for $529.00
Only null left
Add to Cart
 
MS409R1
View Documents
Epredia
MS409R1
Prediluted Unconjugated 1 mL N/A N/A N/A
MS409B
View Documents
Epredia
MS409B
Concentrated Biotin 1 mL N/A N/A N/A
MS409P1
View Documents
Epredia
MS409P1
Concentrated Unconjugated 500 μL N/A N/A N/A
Description

Description

In normal kidney, gp200 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Of other normal tissues examined, the gp200 is also localized along the luminal surfaces of breast lobules and ducts, the luminal surface of the epididymal tubular epithelium, within the cytoplasm of parathyroid parenchymal cells, and focally within the colloid of thyroid follicles. Thirty-one other normal tissues do not express similar or cross-reacting antigens. Reportedly, gp200 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas.

Host Species: Mouse

Clone: PN-15

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Carbohydrate domain

Immunogen: Microsomal fraction of human renal cortical tissue homogenate

Molecular Weight: ∽200kDa

Positive Control: Normal kidney or renal cell CA

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Reduced and nonreduced)
Specifications

Specifications

Renal Cell Carcinoma Marker (gp200) Ab-1
Immunohistochemistry (Paraffin), Western Blot
Mouse
Cancer and Tumor Biology
Primary
PN-15
Monoclonal
IVD
Microsomal fraction of human renal cortical tissue homogenate
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.