Learn More
Trajan Scientific and Medical™ Replacement Needles for Agilent Plunger-in-Needle Syringes
Shop All SGE Inc. Products
Click to view available options
Needle Gauge:
23 G
23 to 26 G
Volume (Metric):
0.5 μL
1 μL
Description
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
Description | Autosampler Syringe Plunger-in-Needle |
Needle Gauge | 23 to 26 G |
Diameter (Metric) Outer Needle | 0.63/0.47 mm |
Volume (Metric) | 0.5 μL |
Length (Metric) Needle | 42 mm |
Needle Point Style | Cone |