Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva RESOURCE™ and SOURCE™ 15 and 5RPC columns

Catalog No. p-4390023
Click to view available options
Volume (Metric):
1.7 mL
10 mL

High Performance Reversed Phase Chromatography

Ideally suited for difficult preparative separations at all scales

  • High resolution separations in minutes
  • High performance at low back pressures
  • High capacity at high flow rates
  • Wide pH stability range
  • Excellent scalability
  • Reproducible quality

TRUSTED_SUSTAINABILITY

Specifications

Ligand Type Matrix surface
Particle Shape Monosized
Particle Size 15 μm
pH 1 to 12
Quantity 10 mL
Volume (Metric) 10 mL
Packing Material Polystyrene/Divinyl Benzene
Endcapped No
Type Reversed Phase Column
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.