Learn More
Restek™ Capillary Installation Gauge for Agilent GCs
SDP
Catalog No. p-3181053 Shop All Restek Corporation Products
Click to view available options
For Use With:
Agilent GC
Varian GCs, 0.0625 in. Ferrules
Seats graphite ferrules onto column for consistent installations.
  • Made from high-quality stainless steel
  • Prevents crushed column ends
  • For 1/16 in. ferrules and fittings
Part No. 21399
  • For Agilent GCs
Part No. 22335
  • For Bruker/Varian GCs
Specifications
Type Gauge
For Use With Varian GCs, 0.0625 in. Ferrules
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.