Learn More
Restek™ Capillary Nuts for Agilent™ 5890/6890/7890 GCs
Shop All Restek Corporation Products
Click to view available options
For Use With (Equipment):
Compact Agilent-style Ferrules
Standard 1/16 in. Ferrules
Material:
Brass
Stainless Steel
Description
- Capillary column nuts
- For use with Agilent GCs
Specifications
Specifications
| Type | Capillary Nut |
| Material | Stainless Steel |
| For Use With (Equipment) | Compact Agilent-style Ferrules |