Learn More
Restek™ FastPack Inlet Kits for Agilent GCs
SDP
Catalog No. 06718410 Shop All Restek Corporation Products
Change view
Click to view available options
Type:
Split w/Wool Inlet Maintenance Kit
Splitless Gooseneck Inlet Maintenance Kit
Splitless Inlet Maintenance Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-718-410 Splitless Inlet Maintenance Kit
06-718-416 Split w/Wool Inlet Maintenance Kit
06-718-413 Splitless Gooseneck Inlet Maintenance Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-718-410 Supplier Restek™ Supplier No. 21101
Only null left
Add to Cart
Add to Cart
Includes all parts needed for routine maintenance of injection ports and the entire system.
  • Economical - costs less than the sum of the individual parts
  • Factory sealed Mylar bag prevents contamination of products from weeks in the lab
  • Each bag includes five maintenance kits
  • Includes: inlet liner (choose from three popular styles), Viton O-ring, 0.8mm ID gold-plated inlet seal and washer, 11mm Thermolite septum
Specifications
Type Splitless Inlet Maintenance Kit
Includes washer, septa and liner
For Use With Agilent GC