Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Pinnacle™ DB Cyano (CN) Mixed Mode HPLC Column, 5 μm
SDP

Catalog No. 06728438 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
100 mm
150 mm
200 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
4.6 mm
Flow Rate:
0.05 mL/min
0.2 mL/min
1 mL/min
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Flow Rate
06-728-438 50 mm 2.1 mm 0.2 mL/min
06-728-417 100 mm 2.1 mm 0.2 mL/min
06-728-421 100 mm 4.6 mm 1 mL/min
06-728-424 200 mm 2.1 mm 0.2 mL/min
06-728-428 200 mm 4.6 mm 1 mL/min
06-728-431 30 mm 2.1 mm 0.2 mL/min
06-728-435 30 mm 4.6 mm 1 mL/min
06-728-442 50 mm 4.6 mm 1 mL/min
06-728-445 150 mm 2.1 mm 0.2 mL/min
06-728-449 150 mm 4.6 mm 1 mL/min
06-728-456 250 mm 4.6 mm 1 mL/min
06-728-452 250 mm 2.1 mm 0.2 mL/min
06-728-444 150 mm 1 mm 0.05 mL/min
06-728-451 250 mm 1 mm 0.05 mL/min
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 06-728-438 Supplier Restek™ Supplier No. 9416552
Add to Cart
Add to Cart

Base-deactivated spherical silica optimized for UHPLC stability.

Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired. It can be used in normal-phase, reversed-phase and HILIC modes. Ideal for analysis of protonated bases.

  • Particle Size: 5μm, spherical
  • Pore Size: 140 angstrom
  • Carbon Load: 4%
  • End-Cap: yes
  • pH Range: 2.5 to 8
  • Temperature Limit: 80°C
  • USP Phase Code: L10
  • Phase Category: Cyano
  • Ligand Type: Cyanopropylsilane

Specifications

Diameter (Metric) 2.1 mm
Film Thickness 5 μm
Flow Rate 0.2 mL/min
For Use With Analysis of a Wide Range of Compounds, From Acidic Through Slightly Basic
Functionality/Interaction Type Electrostatic/Dipole
Length (Metric) 50 mm
Ligand Type Cyanopropylsilane
Particle Shape Spherical
Particle Size 5 μm
pH 2.5 to 8
Pore Size 140 Å
Quantity 5 g
Temperature 80°C
USP Type L10
Stationary Phase Cyano (CN)
Packing Material Silica
Carbon Load 4%
Column Type Mixed Mode
Endcapped Yes
Phase Normal and Reversed Phase
Material Stainless Steel
Technique UHPLC
Product Type Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.