Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Reducing Ferrules: Vespel™/Graphite

Available in graphite or Vespel/graphite material.

Manufacturer:  Restek™ 20215

 View more versions of this product

Catalog No. 06-717-624


  Check Availability
Add to cart

Specifications

Description

  • VG2, 60% Vespel/40% graphite blend offers the best combination of sealing and ease of workability
  • Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
  • Stable to 400°C
  • Recommended for mass spec transfer lines
Description & Specifications

Specifications

Reducing ferrule
0.08cm
Fits Column I.D. 0.53mm
0.317cm
400°C
0.0314 in.
No
0.125 in.
752°F
Graphite/Vespel
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit