Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Rtx-20 GC Capillary Columns
SDP

Catalog No. p-3208149
Click to view available options
Film Thickness:
1 μm
Particle Size:
1 μm

Rtx-20 polymer is synthesized to exacting standards. All residual catalysts and low molecular weight fragments are removed from the polymer, providing a tight monomodal distribution and extremely low bleed.

  • General-purpose columns for volatile compounds, flavor compounds, alcoholic beverages
  • Equivalent to USP G28, G32 phases.

Specifications

Stationary Phase Rtx™-20
Diameter (Metric) Inner 0.53 mm
Type Capillary Column
USP Type G28, G32
Max. Temperature 350°C
For Use With (Application) Volatile Compounds Analysis, Flavor Compounds Analysis, Alcoholic Beverages Analysis
Particle Size 1 μm
Temperature Range -20°C to +260/280°C
Film Thickness 1 μm
Length (Metric) 30 m
Material Stainless Steel
Polarity Low to mid polar
Coating Fused Silica
Quantity 1 Each
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.