Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Rtx™ -5 Capillary Columns - 105m Length
Shop All Restek Corporation Products

Click to view available options
Diameter (Metric) Inner:
0.25 mm
0.32 mm
Film Thickness:
0.25 μm
1 μm
1.0 μm
1.5 μm
3 μm
Particle Size:
0.25 μm
1 μm
1.5 μm
3 μm
Temperature Range:
-60°C to +260/300°C
-60°C to +300/320°C
-60°C to +310/340°C
-60°C to +310/350°C
-60°C to +320/350°C
Description
All residual catalysts and low molecular weight fragments are removed from the Rtx-5 polymer, providing a tight monomodal distribution and extremely low bleed.
- Low-polarity phase
- Crossbond diphenyl dimethyl polysiloxane
- Equivalent to USP G27 and G36 phases
Note
DB-5MS is a silarylene-based polymer, similar to Rxi-5Sil MS.
Order Info
Custom lengths and film thicknesses available.
Specifications
Specifications
| Type | Rtx-5 (G27) Column |
| Diameter (Metric) Inner | 0.25 mm |
| USP Type | G27, G36 |
| Stationary Phase | 5% Diphenyl/95% Dimethyl Polysiloxane |
| Max. Temperature | 350°C |
| For Use With (Application) | Drug Analysis, Solvent Impurities Analysis, Pesticide Analysis, Hydrocarbon Analysis, PCB Congener Analysis, Essential Oils, Semivolatiles Analysis |
| Particle Size | 1 μm |
| Temperature Range | -60°C to +310/340°C |
| Film Thickness | 1.0 μm |
| Length (Metric) | 105 m |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction