Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ RTX™-5 Capillary Columns, 30 m
Shop All Restek Corporation Products

Click to view available options
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
1 μm
1.5 μm
3 μm
5 μm
Includes:
10m Integra-Guard Column
5 in. cage
5m Integra-Guard Column
Cage
Cage, 5m Integra-Guard Column
Integra guard column
Description
- Low polarity phase; Crossbond™ diphenyl dimethyl polysiloxane)
- General-purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners (e.g., Aroclor mixes), essential oils, semivolatiles
- Diphenyl dimethyl polysiloxane stationary phase is the most popular GC stationary phase and is used in a wide variety of applications
- All residual catalysts and low molecular weight fragments are removed from the Rtx™-5 polymer, providing a tight mono-modal distribution and extremely low bleed
- Temperature range: -60°C to 350°C (-76°F to 662°F)
- Equivalent to USP G27 and G36 phases
Specifications
Specifications
Stationary Phase | Rtx™-5 |
USP Type | G27, G36 |
Diameter (Metric) Inner | 0.53 mm |
Max. Temperature | 350°C |
For Use With (Application) | Hydrocarbon Analysis, Solvent Impurities Analysis, Pesticide Analysis, Hydrocarbon Analysis, PCB Congener Analysis, Essential Oils, Semivolatiles Analysis |
Temperature Range | -60°C to +310/330°C |
Film Thickness | 1.5 μm |
Length (Metric) | 30 m |
Material | Stainless Steel |
Polarity | Low polar |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction