Learn More
Restek™ Tubing Dressing Tool
SDP
Catalog No. p-3205690 Shop All Restek Corporation Products
Change view
Click to view available options
Type:
Replacement Needle
Tubing Dressing Tool
Diameter (English) Inner:
1/16 in.
1/8 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Diameter (English) Inner
06-710-543 Replacement Needle 1/16 in.
06-710-545 Replacement Needle 1/8 in.
06-710-544 Tubing Dressing Tool 1/8 in.
06-710-542 Tubing Dressing Tool 1/16 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-710-543 Supplier Restek™ Supplier No. 20189
Only null left
Add to Cart
Add to Cart
Convenient tool opens stainless steel tubing bore and removes burrs.
Tube Dressing Tool is also Included with the SSI TC-20 Tube Cutting Machine.
Specifications
Type Replacement Needle
Diameter (English) Inner 1/16 in.
Packaging Quantity 1 Each