Learn More
Description
Restek Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired. It can be used in normal-phase, reversed-phase and HILIC modes. Ideal for the analysis of protonated bases.
- Particle Size: 5μm, spherical
- Pore Size: 100 angstrom
- Carbon Load: 8%
- End-Cap: yes
- pH Range: 2.5 to 8
- Temperature Limit: 80°C
- USP Phase Code: L10
- Phase Category: cyano
- Ligand Type: cyanopropylsilane
Specifications
Specifications
| Column Chemistry | Nitrile groups |
| Column Format | Preparative Column |
| Diameter (Metric) | 10 mm |
| Film Thickness | 5 μm |
| For Use With | For Normal-Phase, Reversed-Phase (Best with Mobile Phase pH Between 5 and 7), and HILIC Modes |
| Functionality/Interaction Type | Dipole interaction |
| Length (Metric) | 150 mm |
| Ligand Type | Cyanopropylsilane |
| Particle Shape | Spherical |
| Particle Size | 5 μm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
