Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Ultra Cyano HPLC Prep Columns, 5um Particle Size
SDP

Catalog No. 06722383 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
10 mm
21.2 mm
30 mm
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
06-722-383 250 mm 10 mm
06-722-377 100 mm 10 mm
06-722-378 100 mm 21.2 mm
06-722-379 100 mm 30 mm
06-722-374 50 mm 10 mm
06-722-375 50 mm 21.2 mm
06-722-376 50 mm 30 mm
06-722-380 150 mm 10 mm
06-722-381 150 mm 21.2 mm
06-722-382 150 mm 30 mm
06-722-384 250 mm 21.2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 06-722-383 Supplier Restek™ Supplier No. 9106577
Add to Cart
Add to Cart

Easy scale-up from Restek analytical columns.

Restek Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired. It can be used in normal-phase, reversed-phase and HILIC modes. Ideal for the analysis of protonated bases.

  • Particle Size: 5μm, spherical
  • Pore Size: 100 angstrom
  • Carbon Load: 8%
  • End-Cap: yes
  • pH Range: 2.5 to 8
  • Temperature Limit: 80°C
  • USP Phase Code: L10
  • Phase Category: cyano
  • Ligand Type: cyanopropylsilane

Specifications

Column Chemistry Nitrile groups
Column Format Preparative Column
Diameter (Metric) 10 mm
Film Thickness 5 μm
For Use With For Normal-Phase, Reversed-Phase (Best with Mobile Phase pH Between 5 and 7), and HILIC Modes
Functionality/Interaction Type Dipole interaction
Length (Metric) 250 mm
Ligand Type Cyanopropylsilane
Particle Shape Spherical
Particle Size 5 μm
pH 2.5 to 8
Pore Size 100 Å
Surface Area 300 m²/g
Temperature 80°C
USP Type L10
Stationary Phase Cyano (CN)
Packing Material Silica
Carbon Load 8%
Column Type Mixed Mode
Endcapped Yes
Phase Reversed Phase, Normal Phase, HILIC
Technique Reversed or Normal Phase HPLC
Product Type Preparative Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.