Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Male Pipe to Valco™ Internal Adapter Connector from Restek™
SDP

Catalog No. p-3204485 Shop All Restek Corporation Products
Click to view available options
Thread Size:
0.25mm bore
0.75mm bore
Description:
Internal Cross
Internal Tee

For use with 1/16 in. O.D. tubing.

  • 300 series stainless steel
  • Includes stainless steel ferrules
  • Zero-Dead-Volume Valco Internal Tee connects three lines
  • Zero-Dead-Volume Valco Internal Cross connects four lines
  • Union Bore: 0.25mm

Specifications

Type Cross
Diameter (Metric) Inner 0.25 mm
Thread Size 0.25mm bore
Description Internal Cross
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.