Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Viva™ HPLC Columns with C18 Column Packing, 3μm Particle Size

Click to view available options
Length (Metric):
30 mm
50 mm
100 mm
150 mm
Diameter (Metric):
1 mm
2.1 mm
4.6 mm
Flow Rate:
0.07 mL/min
0.31 mL/min
1.5 mL/min
Description
- Rugged, spherical particles with 300 angstrom pore size
- High proportion of pore/surface area available to large molecules
- General-purpose Restek C18 is a conventional monomeric octadecylsilane column
- Particle Size: 3μm
- USP Phase Code: L1
Specifications
Specifications
Diameter (Metric) | 4.6 mm |
Film Thickness | 3 μm |
Flow Rate | 1.5 mL/min |
For Use With | Proteins and Other Higher Molecular Weight Compounds |
Length (Metric) | 100 mm |
Particle Shape | Spherical |
Particle Size | 3 μm |
pH | 2.5 to 10 |
Pore Size | 300 Å |
Temperature | 80°C |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction