Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Restriction Enzyme HaeIII (HC)
Shop All Promega Corporation Products
Click to view available options
Quantity:
12,500 U
Specifications
Specifications
Concentration | 40 to 80 U/μL |
Components | Buffer C 10X Buffer (R003A 1 x 1mL), Bovine Serum Albumin, Acetylated (R396E 1 x 750μL), HaeIII (HC) (R417A1 x 12,500U), MULTI-CORE™ 10X Buffer (R999A 1 x 0.25mL) |
Content And Storage | -20°C |
Source | Haemophilus aegyptius |
Quantity | 12,500 U |
Product Type | Restriction Enzyme HaeIII (HC) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction