Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Restriction Enzyme HpaII
Shop All Promega Corporation Products
Click to view available options
Quantity:
5000 U
Specifications
Specifications
Concentration | 10 U/μL |
Components | Buffer A 10X Buffer (R001A 1 x 1mL), Bovine Serum Albumin, Acetylated (R396E1 x 750μL), HpaII (R631E1 x 5,000U), MULTI-CORE™ 10X Buffer (R999A1 x 0.25mL) |
Content And Storage | -30°C to -10°C |
Source | Haemophilus parainfluenzae |
Quantity | 5000 U |
Product Type | Restriction Enzyme HpaII |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction