Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - Taq I (E)

Catalog No. PRR6151 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
1000 U
Concentration:
10 U/μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
PR-R6151 1000 U 10 U/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PR-R6151 Supplier Promega Supplier No. R6151
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sensitive to overlapping dam methylation. (T/CG A A GC\T)

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 10 U/μL
Components 1 x 1mL 10X Buffer E, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U TaqI, 1 x 0.25mL Multi-Core™ 10X Buffer
Incubator Temperature 37°C
pH 7.4
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 1000 U
Cut Site T.CGA AGC.T
Product Type Taql
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.