Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rex-1/ZFP42 Antibody (5E11E7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP237360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237360 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237360 Supplier Novus Biologicals Supplier No. NBP237360

Mouse Monoclonal Antibody

Rex-1/ZFP42 Monoclonal specifically detects Rex-1/ZFP42 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen Rex-1/ZFP42
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone 5E11E7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ20086, KIAA1584, MGC21637, MGC61687, SUHW4, Suppressor of hairy wing homolog 4, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila)
Gene Symbols ZFP42
Host Species Mouse
Immunogen Purified recombinant fragment of human ZFP42 (AA: 249-310) expressed in E. Coli.
Molecular Weight of Antigen 34.8 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132625
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.