Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Rhyolite Specimen
SCI_ED

Catalog No. S26634 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm

Catalog No. S26634

Supplier: Eisco™ ESNG0049PK10

Only null left
Add to Cart
Add to Cart

Rhyolite Specimen is aphanitic felsic rock, formed during eruptions of thick, viscous magma.

  • For use in density comparison with other igneous rocks
  • Correlates well to Next Generation Science Standards Evidence Statements
  • High silica content makes it difficult for this rock to erupt, leading to explosive eruptions that emit ash and dust into atmosphere
  • Evolved chemistry makes it lightweight relative to other igneous rocks, good identifier during classification exercises

Specifications

Product Type Igneous Rock, Rhyolite
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.