Learn More
Eisco™ Rhyolite Specimen
SCI_ED
Catalog No. p-7076847 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
Rhyolite Specimen is aphanitic felsic rock, formed during eruptions of thick, viscous magma.
  • For use in density comparison with other igneous rocks
  • Correlates well to Next Generation Science Standards Evidence Statements
  • High silica content makes it difficult for this rock to erupt, leading to explosive eruptions that emit ash and dust into atmosphere
  • Evolved chemistry makes it lightweight relative to other igneous rocks, good identifier during classification exercises
Specifications
Product Type Igneous Rock, Rhyolite
For Use With (Application) Density comparison with other igneous rocks
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.