Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROBLD3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB163502 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163502 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163502 Supplier Novus Biologicals Supplier No. NBP281696AF350
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ROBLD3 Polyclonal antibody specifically detects ROBLD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ROBLD3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias ENDAP, Endosomal adaptor protein p14, HSPC003, Late endosomal/lysosomal adaptor and MAPK and MTOR activator 2, late endosomal/lysosomal adaptor, MAPK and MTOR activator 2, Late endosomal/lysosomal Mp1-interacting protein, MAPBPIPRoadblock domain-containing protein 3, MAPKSP1 adaptor protein, MAPKSP1AP, Mitogen-activated protein-binding protein-interacting protein, mitogen-activated protein-binding protein-interacting protein (MAPBPIP), p14Ragulator2, roadblock domain containing 3, ROBLD3
Host Species Rabbit
Immunogen ROBLD3 antibody was raised against an 18 amino acid peptide near the center of human ROBLD3. The immunogen is located within amino acids 40 - 90 of ROBLD3. Amino Acid Squence: WAAYDRNGNQAFNEDNLK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 28956
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.