Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ ROBO1 Pre-design Chimera RNAi
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for all of 3 transcript variants (NM_001145845.1,NM_002941.3,NM_133631.3)
Specifications
Content And Storage | Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. |
---|---|
Species | Human |
For Use With (Application) | RNAi Knockdown |
Includes | DEPC water |
Product Type | ROBO1 Pre-design Chimera RNAi |
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
89-035-712
|
Abnova™
H00006091R03S |
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for all of 3 transcript variants. (NM_001145845.1,NM_002941.3,NM_133631.3) | 10 nmol | N/A | N/A | N/A | |||
89-035-713
|
Abnova™
H00006091R03L |
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for all of 3 transcript variants. (NM_001145845.1,NM_002941.3,NM_133631.3) | 20 nmol | N/A | N/A | N/A | |||
89-035-714
|
Abnova™
H00006091R04S |
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_002941.3,NM_133631.3) | 10 nmol | N/A | N/A | N/A | |||
89-035-715
|
Abnova™
H00006091R04L |
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_002941.3,NM_133631.3) | 20 nmol | N/A | N/A | N/A | |||
89-035-716
|
Abnova™
H00006091R05S |
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_001145845.1, NM_133631.3) | 10 nmol | N/A | N/A | N/A | |||
89-035-717
|
Abnova™
H00006091R05L |
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_001145845.1, NM_133631.3) | 20 nmol | N/A | N/A | N/A | |||
Description
- Gene name: ROBO1
- Gene Alias: DUTT1, FLJ21882, MGC131599, MGC133277, SAX3
- Entrez GeneID: 6091
- Gene Description: Roundabout, axon guidance receptor, homolog 1
Specifications
Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. | |
RNAi Knockdown | |
ROBO1 Pre-design Chimera RNAi |
Human | |
DEPC water | |
Human |
For research use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title