Learn More
Restek™ Roc™ LC Guard Column Cartridges
SDP
Catalog No. 06611535 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With:
Roc Phenyl-Hexyl HPLC Columns
Roc Silica HPLC Columns
Roc™ C18 HPLC Columns
Roc™ C8 HPLC Columns
Roc™ Cyano HPLC Columns
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
06-611-535 Roc Silica HPLC Columns
06-611-536 Roc™ C8 HPLC Columns
06-611-537 Roc™ C18 HPLC Columns
06-611-539 Roc™ Cyano HPLC Columns
06-611-538 Roc Phenyl-Hexyl HPLC Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 06-611-535 Supplier Restek™ Supplier No. 953050210
Add to Cart
Add to Cart
Protects Roc™ LC Columns with minimal effect on retention, peak shape, or efficiency
  • Guard Column Cartridges require Roc™ Guard Column Holder (cat. #25812)
Specifications
Diameter (Metric) 4 mm
For Use With Roc Silica HPLC Columns
Length (Metric) 10 mm
Quantity 3/Pk.
Product Type Guard Cartridge