Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Safe-Lock Tubes 1.5 mL - Microtube
GSA_VAGREENER_CHOICE

Catalog No. e0030123611 Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Amber
Assorted
Blue
Clear
Green
Red
Yellow
Purity or Quality Grade:
Biopur™
Eppendorf Quality
Forensic DNA Grade
PCR Clean
Max. RCF:
22,000 x g
30,000 x g
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Purity or Quality Grade Max. RCF
E0030123611 Clear Forensic DNA Grade 22,000 x g
05-402-27 Blue Eppendorf Quality 30,000 x g
05-402-28 Green Eppendorf Quality 30,000 x g
05-402-29 Red Eppendorf Quality 30,000 x g
05-402-30 Yellow Eppendorf Quality 30,000 x g
05-402-25 Clear Eppendorf Quality 30,000 x g
05-402-5 Assorted Eppendorf Quality 30,000 x g
05-402-94 Clear PCR Clean 30,000 x g
05-402-26 Amber PCR Clean 30,000 x g
05-402-24B Clear Biopur™ 22,000 x g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. E0030123611 Supplier Eppendorf™ Supplier No. 0030123611
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Eppendorf™ Safe-Lock Tubes 1.5 mL - Microtube ensures secure sample storage. With a patented Safe-Lock design, these microtubes prevent accidental opening, safeguarding samples. Ideal for diverse applications, they offer reliability, durability, and ease of use in a 1.5 mL capacity.

  • The hinged Eppendorf Safe-Lock lid prevents inadvertent lid opening during incubation and storage.
  • g-Safe™ technology, depending on the Safe-Lock Tube variant (refer to the manual), ensures outstanding centrifugation stability up to 30,000 x g, preventing sample loss due to tube breakage and providing additional safety when handling hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances demonstrated to leach from plastic consumables into the sample and adversely impact bio-assay results.
  • Polypropylene construction offers robust resistance to chemicals, mechanical stress, and extreme temperatures.
  • Precision lid sealing minimizes evaporation rates during extended storage periods.
  • The generously sized frosted lid and side surface facilitate easy labeling.
  • Assured functionality in a temperature range from -86 °C to 100 °C.
  • Autoclavable when open (121 °C, 20 min) for reliable sterilization.
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur™ quality for tested purity.

Order Info

Supplied individually wrapped in your choice of three sizes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Cap Style Hinged lid
Closure Color Colorless
Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Clear
Material Polypropylene
For Use With (Application) Incubation and storage
No. per Pack 500 Pk.
Packaging 500 Tubes (10 Bags x 50 Tubes)
Temperature Range (English) -122.8°F to +212°F
Temperature Range (Metric) -86°C to +100°C
Sterility Non-sterile
Purity or Quality Grade Forensic DNA Grade
Graduated Yes
Capacity (English) 0.05 oz.
Closure Type Safe-lock Lid
Max. RCF 22,000 x g
Shape Cylindrical
Product Type Locking Snap-cap Microtube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.