Description
- For use in studying depositional environments
- Correlates well to Next Generation Science Standards Evidence Statements
- Rounded, well-sorted sand grains are excellent identifiers for students during sedimentary rock identification
- This indicates that clasts in rocks have traveled long distance from their source and were heavily eroded along way
Specifications
Specifications
| Product Type | Sedimentary Rock, Sandstone |
| For Use With (Application) | Studying depositional environments |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 1 Ea. |
| Specimen Size | 2 to 3 cm |