Learn More
SARS-CoV-2 Nucleocapsid Antibody (D207) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | SARS-CoV-2 Nucleocapsid |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 - 1:2000, ELISA 1:5000 - 1:10000, Sandwich ELISA |
| Formulation | PBS (pH 7.5) |
| Gene Alias | 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein |
| Host Species | Human |
| Immunogen | Recombinant SARS-CoV-2 Nucleocapsid protein |
| Purification Method | >95% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.