Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Nucleocapsid Protein Antibody, Novus Biologicals™
SDP

Catalog No. NB10056049T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056049T 0.025 mg
NB10056049 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056049T Supplier Novus Biologicals Supplier No. NB10056049SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

SARS Nucleocapsid Protein Polyclonal specifically detects SARS Nucleocapsid Protein in SARS-CoV samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SARS Nucleocapsid Protein
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation PBS containing 0.5% BSA with 0.05% Sodium Azide
Gene Accession No. NP_006504
Gene Alias COVID-19 nucleocapsid protein, N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, SARSCOV2 N protein, SARS-COV-2 nucleocapsid protein, Severe acute respiratory syndrome
Host Species Rabbit
Immunogen The antibody was developed by immunizing Rabbit with a synthetic peptide corresponding to amino acids 84-99 (C-IGYYRRATRRIRGGDG) from the N (SARS Nucleocapsid) for the Human SARS coronavirus (Genbank accession no. YP_009724397.2)
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6301
Target Species SARS-CoV
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.