Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SB 203580, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
SB 203580 is a selective inhibitor of p38 MAPK (IC50 values are 50 and 500 nM for SAPK2a/p38 and SAPK2b/p38β2 respectively). Displays 100-500-fold selectivity over LCK, GSK-3β and PKBα. Shown to inhibit IL-2-induced T cell proliferation, cyclooxygenase-1 and -2, and thromboxane synthase. Enhances clonal growth of skin epithelial progenitor cells; stimulates neural stem cell (NSC) proliferation. SB 203580 is an essential component of medium for maintaining stem cells in naive pluripotent state and can be used to promote expansion of HSCs ex vivo.
Water-soluble Salt also available.
Specifications
Quantity | 1 mg |
Formula Weight | 377.44 |
Percent Purity | >98% |
Chemical Name or Material | SB 203580 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction