Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SB 258719 hydrochloride
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective 5-HT7 receptor antagonist that displays > 100-fold selectivity over a range of other receptors. Reverses the hypothermic effect of 5-CT in mice following i.p. administration.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 374.97 |
Purity | >99% |
Molecular Weight (g/mol) | 374.968 |
Chemical Name or Material | SB 258719 hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction