Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Scoria Specimen
SCI_ED

Catalog No. S26614 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm

Catalog No. S26614

Supplier: Eisco™ ESNG0050PK10

Only null left
Add to Cart
Add to Cart

Scoria forms when spatter from bubbly mafic lava cools quickly.

  • For use in lessons on eruption style and cooling rate
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Erupted from spatter cone as large air bubbles made their way through magma to surface
  • Crystallization happens so quickly that large air pockets are trapped within rock, often giving samples jagged, sharp appearance

Specifications

Product Type Igneous Rock, Scoria
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.