Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | SCP1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot reported in scientific literature (PMID 23843895), Simple Western 1:200, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:300-1:750, Immunohistochemistry-Paraffin reported in scientific literature (PMID 26858254), Immunoblotting reported in scientific literature (PMID 27431324) |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300228SS
![]() |
Novus Biologicals
NB300228SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB300228
![]() |
Novus Biologicals
NB300228 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
SCP1 Polyclonal specifically detects SCP1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
SCP1 | |
Western Blot reported in scientific literature (PMID 23843895), Simple Western 1:200, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:300-1:750, Immunohistochemistry-Paraffin reported in scientific literature (PMID 26858254), Immunoblotting reported in scientific literature (PMID 27431324) | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1 | |
SYCP1 | |
IgG | |
Affinity Purified |
1 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Primate | |
Q15431 | |
6847 | |
A C-terminal synthetic peptide made to the human SCP1 protein sequence. [UniProt# Q15431] | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title