Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDE2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | C1orf55 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15637320
![]() |
Novus Biologicals
NBP15637320UL |
20 μL |
Each for $158.00
|
|
|||||
NBP156373
![]() |
Novus Biologicals
NBP156373 |
100 μL |
Each for $487.50
|
|
|||||
Description
SDE2 Polyclonal specifically detects SDE2 in Human samples. It is validated for Western Blot.Specifications
C1orf55 | |
Polyclonal | |
Rabbit | |
Q6IQ49 | |
163859 | |
Synthetic peptide directed towards the C terminal of human C1orf55 (NP_689821). Peptide sequence AFTSVAELELLGLEKLKCELMALGLKCGGTLQERAARLFSVRGLAKEQID. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
C1orf55, chromosome 1 open reading frame 55, dJ671D7.1, FLJ35382, hypothetical protein LOC163859, RP4-671D7.1, SDE2 telomere maintenance homolog (S. pombe) | |
SDE2 | |
IgG | |
50 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title