Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Seasonal H1N1 Nucleocapsid Protein Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-200074552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396501 0.025 mg
NBP241114 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396501 Supplier Novus Biologicals Supplier No. NBP2411140.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Seasonal H1N1 Nucleocapsid Protein Polyclonal specifically detects Seasonal H1N1 Nucleocapsid Protein in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.

Specifications

Antigen Seasonal H1N1 Nucleocapsid Protein
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias Nucleocapsid protein, Nucleoprotein, Protein N
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide from the seasonal H1N1 NP protein. The immunogen is located within amino acids 350 - 400 of Seasonal H1N1 Nucleocapsid Protein. Amino Acid Squence: NMDNMGSS
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific for the seasonal H1N1 influenza NP and will not recognize the corresponding NP sequence from the swine-origin H1N1 influenza (A/California/14/2009 (H1N1)).
Target Species Influenza virus, Virus
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.