Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEPN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | SEPN1 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16905820
![]() |
Novus Biologicals
NBP16905820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP169058
![]() |
Novus Biologicals
NBP169058 |
100 μL |
Each for $487.50
|
|
|||||
Description
SEPN1 Polyclonal specifically detects SEPN1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SEPN1 | |
Unconjugated | |
RUO | |
FLJ24021, rigid spine muscular dystrophy 1, RSMD1, RSS, selenoprotein N, selenoprotein N, 1, SelN, SELNMDRS1 | |
SEPN1 | |
IgG | |
62 kDa |
Polyclonal | |
Rabbit | |
D3Z5N3 | |
57190 | |
Synthetic peptides corresponding to Sepn1 (selenoprotein N, 1) The peptide sequence was selected from the C terminal of Sepn1. Peptide sequence VHHINANYFLDITSMKPEDMENNNVFSFSSSFEDPSTATYMQFLREGLRR. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title