Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | SETD4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18004520
![]() |
Novus Biologicals
NBP18004520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180045
![]() |
Novus Biologicals
NBP180045 |
100 μL |
Each for $487.50
|
|
|||||
Description
SETD4 Polyclonal specifically detects SETD4 in Human samples. It is validated for Western Blot.Specifications
SETD4 | |
Polyclonal | |
Rabbit | |
C21orf18, C21orf27, chromosome 21 open reading frame 18, chromosome 21 open reading frame 27, SET domain containing 4, SET domain-containing protein 4 | |
SETD4 | |
IgG | |
32 kDa |
Western Blot | |
Unconjugated | |
RUO | |
54093 | |
Synthetic peptide directed towards the C terminal of human SETD4. Peptide sequence VQVKAAFNEETHSYEIRTTSRWRKHEEVFICYGPHDNQRLFLEYGFVSVH. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title