Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SF3B4 Antibody (3A1), Novus Biologicals™
SDP

Catalog No. NBP192692SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP192692SS 0.025 mL
NBP192692 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP192692SS Supplier Novus Biologicals Supplier No. NBP192692SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

SF3B4 Monoclonal specifically detects SF3B4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SF3B4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3A1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Accession No. Q15427
Gene Alias Hsh49, pre-mRNA splicing factor SF3b 49 kDa subunit, Pre-mRNA-splicing factor SF3b 49 kDa subunit, SAP 49, SAP49MGC10828, SF3B49, SF3b50, spliceosomal protein, spliceosome-associated protein (U2 snRNP), Spliceosome-associated protein 49, splicing factor 3B subunit 4, splicing factor 3b, subunit 4, 49kD, splicing factor 3b, subunit 4, 49kDa
Gene Symbols SF3B4
Host Species Mouse
Immunogen Full length recombinant human SF3B4 expressed in and purified from E. coli. [UniProt# Q15427]
Molecular Weight of Antigen 49 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10262
Test Specificity Human, Bovine, Porcine, Mouse, Rat and other Mammals.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.