Learn More
Trajan™ GC/MS Interface Connections for Varian GC Instruments
Shop All SGE Inc. Products
Click to view available options
For Use With:
0.10 to 0.25 mm Column
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Specifications
Type | Ferrule |
Packaging Quantity | 10 ferrules, 2 nuts |
For Use With | 0.10 to 0.25 mm Column |
Material | SilTite |